Learn More
Aviva Systems Biology Corp CXCL6 Antibody - middle region (ARP30969_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP30969_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CXCL6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CXCL6. Alternate names: CKA-3, GCP-2, GCP2, SCYB6ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Rabbit, Rat
Catalog No. 50-014-54173