Learn More
Novus Biologicals™ CXCL8/IL-8 ELISA Development Kit

Specifications
Specifications
| Assay Range | 8 - 800 pg/mL |
| Product Type | ELISA Development Kit |
| Sample Type | Serum, Plasma, Cell Culture Supernatant |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 3576 |
| Gene Symbol | CXCL8 |
| Quantity | 1 Kit (for 6 Plates) |
| Research Discipline | Angiogenesis, Biologically Active Proteins, Cancer, Chemokines and Cytokines, Cytokine Research, Immunology, Innate Immunity, Signal Transduction |
| Storage Requirements | Storage of components varies. See protocol for specific instructions. |
| Synonym | 3-10°C, AMCF-I, C-X-C motif chemokine 8, CXCL8SCYB8, Emoctakin, GCP-1TSG-1, interleukin 8, K60, LECT, MDNCFb-ENAP, member 8, MONAPGCP1, NAP-1NAP1, Neutrophil-activating protein 1, Protein 3-10°C, T cell chemotactic factor, T-cell chemotactic factor |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.