Learn More
Biomatik Corporation Cxcl8 Monoclonal Antibody
Supplier:  Biomatik Corporation CAC10258
Encompass_Preferred
Cxcl8 Monoclonal Antibody is a Human (Homo sapiens) Monoclonal Antibody to CXCL8. The Cxcl8 Monoclonal Antibody has been validated for the following applications: ELISA, IHC. Host: Mouse. Species: Human (Homo sapiens). Immunogen: Recombinant Human Interleukin-8 Protein. Applications: ELISA, IHC. Target Synonyms: CXCL8. Accession Number: P10145. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5467