Learn More
Aviva Systems Biology Corp CXCL9 Antibody - middle region (AVARP07022_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP07022_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CXCL9. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CXCL9. Alternate names: CMK, Humig, MIG, SCYB9, crg-10ApplicationWB. Species reactivity: Cow, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-62545