Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Prilocaine Hydrochloride, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18605813 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-813 5 kg Amber Glass Bottle
18-605-809 25 g Amber Glass Bottle
18-605-810 100 g Amber Glass Bottle
18-605-811 500 g Amber Glass Bottle
18-605-812 1 kg Amber Glass Bottle
18-605-808 5 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-605-813 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. PR1025KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

PR102, 1786-81-8, C13H20N2O·HCl

Spectrum™ Chemical Prilocaine Hydrochloride, USP is a local anesthetic used most often for infiltration anesthesia in dentistry. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1786-81-8
Molecular Formula C13H21ClN2O
Molecular Weight (g/mol) 256.77
InChI Key BJPJNTKRKALCPP-UHFFFAOYNA-N
IUPAC Name hydrogen N-(2-methylphenyl)-2-(propylamino)propanamide chloride
SMILES [H+].[Cl-].CCCNC(C)C(=O)NC1=CC=CC=C1C

Specifications

CAS Max % 1
Ignition Residue 0.001
Loss on Drying 0.003
Packaging Amber Glass Bottle
Quantity 5 kg
Percent Purity 98 to 102%
Grade USP
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Prilocaine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.