Learn More
MilliporeSigma CXCR2 MEMBRANE PREPARATION
Supplier:  MilliporeSigma HTS002M
Encompass
Chemiscreen Membrane Preparation Human Recombinant CXCR2 Chemokine Receptor, 200 tests, For Radioligand binding assay, Reacts with Human, Maintain frozen at -70 deg.C for up to 2 years
Catalog No. 50-818-528