Learn More
Eurofins Discoverx ChemiBrite CXCR3 Chemokine Receptor Stable Cell Line, 2 vials. (HTS003L)
Catalog No. 502005347
Encompass
The ChemiBrite CXCR3 Chemokine Receptor cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand based activation of CXCR3 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses CXCR3 alongside endogenous high levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. ChemiBrite cells also express a clytin variant, allowing for luminescent as well as fluorescent signal detection. Upon activation of CXCR3, the resulting calcium activity can be assayed in either detection mode. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, visit www.discoverx.com/targets/gpcr-target-biology.
Catalog No. 50-200-5347 Supplier Eurofins Discoverx Supplier No. HTS003L
Add to Cart
Add to Cart
Request a Quote
The ChemiBrite CXCR3 Chemokine Receptor cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand based activation of CXCR3 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses CXCR3 alongside endogenous high levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. ChemiBrite cells also express a clytin variant, allowing for luminescent as well as fluorescent signal detection. Upon activation of CXCR3, the resulting calcium activity can be assayed in either detection mode. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, visit www.discoverx.com/targets/gpcr-target-biology.