Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cxcr3 protein, human (cell-free, C-His tag) | >80.0% | 10UG
SDP

Catalog No. 5000176480
Encompass_Preferred

Recombinant human C-X-C chemokine receptor 3 (CXCR3) protein with a C-terminal His tag, expressed in Escherichia coli and provided as a purified recombinant reagent for research use. The sequence corresponds to UniProt P49682 (368 amino acids, ~40,660 Da). Certificate of analysis reports purity greater than 80% by reducing SDS-PAGE.

  • Recombinant full-length human CXCR3 expressed in E. coli.
  • C-terminal His tag for affinity purification and detection.
  • Sequence identity referenced to UniProt accession P49682.
  • Purity greater than 80% as determined by reducing SDS-PAGE.
  • Intended for research use only.

Catalog No. 50-001-76480 Supplier Medchemexpress LLC Supplier No. HYP700539A10UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human C-X-C chemokine receptor 3 (CXCR3) protein with a C-terminal His tag, expressed in Escherichia coli and provided as a purified recombinant reagent for research use. The sequence corresponds to UniProt P49682 (368 amino acids, ~40,660 Da). Certificate of analysis reports purity greater than 80% by reducing SDS-PAGE.

  • Recombinant full-length human CXCR3 expressed in E. coli.
  • C-terminal His tag for affinity purification and detection.
  • Sequence identity referenced to UniProt accession P49682.
  • Purity greater than 80% as determined by reducing SDS-PAGE.
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.