Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CXCR4 Human N-His- 10ug
SDP

Catalog No. 5000183225
Encompass_Preferred

Recombinant human CXCR4 fragment (residues P303-S352) expressed in E. coli and provided with N-terminal His and SUMO tags and a C-terminal Myc tag. The protein is supplied as a purified fragment (~28 kDa, >90% purity) intended for biochemical and cell-based studies of CXCR4-CXCL12 interactions, receptor signaling, and protein-protein interaction assays.

  • Expressed in E. coli for consistent recombinant production.
  • Contains N-terminal His and SUMO tags and a C-terminal Myc tag.
  • Fragment corresponds to residues P303-S352 of human CXCR4.
  • Approximately 28 kDa molecular weight.
  • Purity greater than 90% by reducing SDS-PAGE.
  • Supplied in 10 μg size suitable for assay development and binding studies.

Catalog No. 50-001-83225 Supplier Medchemexpress LLC Supplier No. HYP7215910UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human CXCR4 fragment (residues P303-S352) expressed in E. coli and provided with N-terminal His and SUMO tags and a C-terminal Myc tag. The protein is supplied as a purified fragment (~28 kDa, >90% purity) intended for biochemical and cell-based studies of CXCR4-CXCL12 interactions, receptor signaling, and protein-protein interaction assays.

  • Expressed in E. coli for consistent recombinant production.
  • Contains N-terminal His and SUMO tags and a C-terminal Myc tag.
  • Fragment corresponds to residues P303-S352 of human CXCR4.
  • Approximately 28 kDa molecular weight.
  • Purity greater than 90% by reducing SDS-PAGE.
  • Supplied in 10 μg size suitable for assay development and binding studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.