Learn More
Enzo Life Sciences CXCR4 (mouse) polyclonal antibody (200µg)
SDP
Catalog No. 502008166
Encompass_Preferred
Host: Goat. Immunogen: Synthetic peptide corresponding to aa 14-40 (Y14SEEVGSGDYDS NKEPCFRDENVHFNR40) of the N-terminal extracellular domain of mouse CXCR4 (CXC chemokine receptor 4). Species reactivity: Mouse. Applications: ICC, IHC (PS), WB. Recommended Dilutions: Immunocytochemistry (1:300) Immunohistochemistry (paraffin sections, 1:200) Western Blot (1:1,000) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Epitope-affinity purified IgG. Formulation: Liquid. In PBS containing 1mg/ml BSA and 0.1% sodium azide. Handling: For maximum product recovery after thawing, centrifuge the vial before opening the cap. Avoid freeze/thaw cycles.
Catalog No. 50-200-8166 Supplier Enzo Life Sciences Supplier No. ALX210819C200
Host: Goat. Immunogen: Synthetic peptide corresponding to aa 14-40 (Y14SEEVGSGDYDS NKEPCFRDENVHFNR40) of the N-terminal extracellular domain of mouse CXCR4 (CXC chemokine receptor 4). Species reactivity: Mouse. Applications: ICC, IHC (PS), WB. Recommended Dilutions: Immunocytochemistry (1:300) Immunohistochemistry (paraffin sections, 1:200) Western Blot (1:1,000) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Epitope-affinity purified IgG. Formulation: Liquid. In PBS containing 1mg/ml BSA and 0.1% sodium azide. Handling: For maximum product recovery after thawing, centrifuge the vial before opening the cap. Avoid freeze/thaw cycles.