Learn More
Eurofins Discoverx Ready-to-Assay CXCR5 Chemokine Receptor Frozen Cells, 2 Vials of Cells. (HTS055RTA)
Catalog No. 502005762
Encompass
The Ready-to-Assay CXCR5 Chemokine Receptor Frozen Cells are a simple, thaw-and-assay cell format for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This assay assesses ligand (e.g. CXCL13/BCA-1/BLC) based activation of CXCR5 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses CXCR5 alongside exogenous levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. Upon activation of CXCR5, the resulting calcium activity can be fluorescently assayed. Ready-to-assay kits consist of two vials of the stably modified clonal cell line in a convenient thaw-and-use format. For detailed product information, User manual and datasheet, visit www.discoverx.com/products-applications/frozen-cells.
Catalog No. 50-200-5762 Supplier Eurofins Discoverx Supplier No. HTS055RTA
The Ready-to-Assay CXCR5 Chemokine Receptor Frozen Cells are a simple, thaw-and-assay cell format for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This assay assesses ligand (e.g. CXCL13/BCA-1/BLC) based activation of CXCR5 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses CXCR5 alongside exogenous levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. Upon activation of CXCR5, the resulting calcium activity can be fluorescently assayed. Ready-to-assay kits consist of two vials of the stably modified clonal cell line in a convenient thaw-and-use format. For detailed product information, User manual and datasheet, visit www.discoverx.com/products-applications/frozen-cells.