Learn More
Medchemexpress LLC Cxcr7 modulator 1 | 2231812-31-8 | 99.5% | 914.07 | C48H57F2N7O7S | 50 MG
SDP
Catalog No. 5000242783
Encompass_Preferred
CXCR7 modulator 1 is a potent, orally bioavailable peptoid-hybrid modulator of the chemokine receptor CXCR7 with an in vitro Ki of 9 nM. It is supplied for preclinical biochemical and pharmacological research into CXCR7 signaling and ligand-receptor interactions.
  • Potent CXCR7 affinity (Ki = 9 nM)
  • Orally bioavailable chemical scaffold suitable for in vivo dosing studies
  • High reported purity for reliable experimental results
  • Supplied as a solid suitable for formulation and analytical work
  • Available in multiple small research sizes for dose-ranging studies
Catalog No. 50-002-42783 Supplier Medchemexpress LLC Supplier No. HY10798750MG
CXCR7 modulator 1 is a potent, orally bioavailable peptoid-hybrid modulator of the chemokine receptor CXCR7 with an in vitro Ki of 9 nM. It is supplied for preclinical biochemical and pharmacological research into CXCR7 signaling and ligand-receptor interactions.
  • Potent CXCR7 affinity (Ki = 9 nM)
  • Orally bioavailable chemical scaffold suitable for in vivo dosing studies
  • High reported purity for reliable experimental results
  • Supplied as a solid suitable for formulation and analytical work
  • Available in multiple small research sizes for dose-ranging studies