Learn More
Medchemexpress LLC CXCR7 modulator 1 | 2231812-31-8 | 99.5% | 914.07 g/mol | C48H57F2N7O7S | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10798725MG
Encompass_Preferred
CXCR7 modulator 1 is a potent, orally bioavailable peptoid-hybrid modulator of the chemokine receptor CXCR7 with a reported Ki of 9 nM. Supplied as a solid research compound, it is intended for biochemical and cellular studies and is typically dissolved in DMSO for in vitro or in vivo formulation.
Catalog No. 50-002-42452