Learn More
Medchemexpress LLC CXCR7 modulator 1 (compound 25) | 2231812-31-8 | 99.5% | 914.07 | C48H57F2N7O7S | 100 MG
SDP
Catalog No. 5000242220
Encompass_Preferred
CXCR7 modulator 1 is a potent, orally bioavailable peptoid-hybrid small molecule that modulates the chemokine receptor CXCR7 (reported Ki = 9 nM). It is intended for research use in studies of GPCR signaling and immunology/inflammation.
  • Potent CXCR7 modulator with reported Ki of 9 nM.
  • Orally bioavailable peptoid-hybrid small molecule.
  • Peptoid-hybrid scaffold suitable for medicinal chemistry studies.
  • High purity (99.5%) for research applications.
  • Molecular weight 914.07, molecular formula C48H57F2N7O7S.
  • Suitable for GPCR signaling and immunology or inflammation research.
Catalog No. 50-002-42220 Supplier Medchemexpress LLC Supplier No. HY107987100MG
CXCR7 modulator 1 is a potent, orally bioavailable peptoid-hybrid small molecule that modulates the chemokine receptor CXCR7 (reported Ki = 9 nM). It is intended for research use in studies of GPCR signaling and immunology/inflammation.
  • Potent CXCR7 modulator with reported Ki of 9 nM.
  • Orally bioavailable peptoid-hybrid small molecule.
  • Peptoid-hybrid scaffold suitable for medicinal chemistry studies.
  • High purity (99.5%) for research applications.
  • Molecular weight 914.07, molecular formula C48H57F2N7O7S.
  • Suitable for GPCR signaling and immunology or inflammation research.