Learn More
Medchemexpress LLC HY-112154 10mg Medchemexpress, CXCR7 modulator 2 CAS: Purity:>98%
SDP
Catalog No. 501969040
Encompass_Preferred
Medchemexpress, HY-112154 10mg CXCR7 modulator 2 CAS: CXCR7 modulator 2 is a modulator of C-X-C Chemokine Receptor Type 7 (CXCR7), with a Ki of 13 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9040 Supplier Medchemexpress LLC Supplier No. HY11215410MG
Medchemexpress, HY-112154 10mg CXCR7 modulator 2 CAS: CXCR7 modulator 2 is a modulator of C-X-C Chemokine Receptor Type 7 (CXCR7), with a Ki of 13 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.