Learn More
Medchemexpress LLC CXCR7 modulator 2 | 2227426-37-9 | 98.7% | 522.68 | 50 MG
SDP
Catalog No. 5000436168
Encompass_Preferred
CXCR7 modulator 2 is a C-X-C Chemokine Receptor Type 7 (CXCR7) modulator with a Ki of 13 nM. It is intended for research use only.
  • Potent CXCR7-binding affinity (Ki=13 nM)
  • Demonstrates β-arrestin activity (EC50=11 nM)
  • Improved selectivity in GPCR panel
  • Improved therapeutic index in hERG patch-clamp assay
  • Moderate to high in vitro turnover in mouse-liver microsomes and hepatocytes
  • Good aqueous solubility
  • Significantly reduced cardiac fibrosis in isoproterenol-induced cardiac injury model in mice
Catalog No. 50-004-36168 Supplier Medchemexpress LLC Supplier No. HY11215450MG
CXCR7 modulator 2 is a C-X-C Chemokine Receptor Type 7 (CXCR7) modulator with a Ki of 13 nM. It is intended for research use only.
  • Potent CXCR7-binding affinity (Ki=13 nM)
  • Demonstrates β-arrestin activity (EC50=11 nM)
  • Improved selectivity in GPCR panel
  • Improved therapeutic index in hERG patch-clamp assay
  • Moderate to high in vitro turnover in mouse-liver microsomes and hepatocytes
  • Good aqueous solubility
  • Significantly reduced cardiac fibrosis in isoproterenol-induced cardiac injury model in mice