Learn More
Medchemexpress LLC HY-112154 5mg Medchemexpress, CXCR7 modulator 2 CAS: Purity:>98%
SDP
Catalog No. 501932416
Encompass_Preferred
Medchemexpress, HY-112154 5mg CXCR7 modulator 2 CAS: CXCR7 modulator 2 is a modulator of C-X-C Chemokine Receptor Type 7 ( CXCR7 ), with a K i of 13 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2416 Supplier Medchemexpress LLC Supplier No. HY1121545MG
Medchemexpress, HY-112154 5mg CXCR7 modulator 2 CAS: CXCR7 modulator 2 is a modulator of C-X-C Chemokine Receptor Type 7 ( CXCR7 ), with a K i of 13 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.