Learn More
Aviva Systems Biology Corp CXorf20 Antibody - middle region (ARP53161_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53161_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CXorf20. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CXorf20. Alternate names: MGC33653, CXorf20ApplicationWB. Species reactivity: Human
Catalog No. 50-014-60950