Learn More
Aviva Systems Biology Corp CXorf34 Antibody - middle region (ARP48928_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48928_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CXorf34. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CXorf34. Alternate names: FLJ12687, dJ341D10.3, CXorf34ApplicationWB. Species reactivity: Dog, Horse, Human, Mouse, Pig, Rabbit
Catalog No. 50-014-53565