Learn More
Aviva Systems Biology Corp CXORF6 Antibody - N-terminal region (ARP34221_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34221_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CXORF6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CXORF6. Alternate names: CG1, F18, HYSP2, CXorf6ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-55306