Learn More
Aviva Systems Biology Corp CXorf67 Antibody - middle region (ARP55962_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55962_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LOC340602. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human LOC340602. Alternate names: MGC47837ApplicationWB. Species reactivity: Dog, Human
Catalog No. 50-286-100