Learn More
Aviva Systems Biology Corp CXorf9 Antibody - N-terminal region (ARP57305_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57305_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CXorf9. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CXorf9. Alternate names: 753P9, SLY, HACS2, CXorf9, SH3D6CApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-62124