Learn More
Labrepco Inc CXR100 SHIPPER WITH REPLACEABL
SDP
Supplier:  Labrepco Inc CXR10011M
Encompass_Preferred
Shipper, Dry; Labrepco; Cryo Express 100; w/replaceable adsorbent ; w/11 in. canister ; IATA compliant; Magniformed necktube; Aluminum; w/lightweight case; Wt. (Empty): 11.7 lb. (5.3kg); 4.4L liquid nitrogen adsorbed; 18.4 in. H (467mm)
Catalog No. 50-902-0752