Learn More
Taylor Wharton CXR500 Cryogenic Shipper with Replaceable Absorbent Material without Rack (Capacity: 500 x 2.0mL Vials)
Catalog No. NC0053916
Encompass_Preferred
Cryo Express (CXR500) dry shipper designed to safely transport a variety of materials at cryogenic temperatures with replaceable absorbent. Complies with IATA regulations.
Catalog No. NC0053916 Supplier Taylor Wharton Supplier No. CXR5000
Cryo Express (CXR500) dry shipper designed to safely transport a variety of materials at cryogenic temperatures with replaceable absorbent. Complies with IATA regulations.