Learn More
Aviva Systems Biology Corp CXXC1 Antibody - C-terminal region (ARP39110_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39110_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CXXC1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human CXXC1. Alternate names: 2410002I16Rik, 5830420C16Rik, CGBP, HsT2645, PCCX1, PHF18, hCGBP, CFP1, SPP1, ZCGPC1ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53126