Learn More
Aviva Systems Biology Corp Cxxc1 Antibody - N-terminal region (ARP43590_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43590_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Cxxc1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N-terminal region of Rat Cxxc1. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Goat, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-084