Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cy-09 | 1073612-91-5 | MFCD31619349 | 99.6% | 423.43 g/mol | C19H12F3NO3S2 | 1 ML
SDP

Catalog No. 5000246863
Encompass_Preferred

CY-09 is a small-molecule inhibitor of the NLRP3 inflammasome (CAS 1073612-91-5) supplied as a ready-to-use 10 mM solution in DMSO for research applications. It binds the ATP-binding motif of the NLRP3 NACHT domain to inhibit ATPase activity and suppress inflammasome assembly and activation. For research use only.

  • Ready-to-use 10 mM solution in DMSO.
  • Supplied volume 1 mL per vial.
  • High reported purity (99.58%).
  • Molecular weight 423.43 g/mol; formula C19H12F3NO3S2.
  • Certificate of analysis and safety data sheet available.

Catalog No. 50-002-46863 Supplier Medchemexpress LLC Supplier No. HY10366610MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CY-09 is a small-molecule inhibitor of the NLRP3 inflammasome (CAS 1073612-91-5) supplied as a ready-to-use 10 mM solution in DMSO for research applications. It binds the ATP-binding motif of the NLRP3 NACHT domain to inhibit ATPase activity and suppress inflammasome assembly and activation. For research use only.

  • Ready-to-use 10 mM solution in DMSO.
  • Supplied volume 1 mL per vial.
  • High reported purity (99.58%).
  • Molecular weight 423.43 g/mol; formula C19H12F3NO3S2.
  • Certificate of analysis and safety data sheet available.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.