Learn More
Medchemexpress LLC HY-106094 10mg Medchemexpress, CY 208-243 CAS:100999-26-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10609410MG
Encompass_Preferred
Medchemexpress, HY-106094 10mg CY 208-243 CAS:100999-26-6 CY 208-243 is a selective dopamine D1 receptor agonist which exhibits antiparkinsonian activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8317