Learn More
Medchemexpress LLC Cy 5 amine TFA | 99.8% | 731.33 g/mol | C40H54ClF3N4O3 | 10 MG
SDP
Catalog No. 5000297524
Encompass_Preferred
Cy 5 amine TFA is a cyanine fluorescent amine supplied as the trifluoroacetic acid (TFA) salt. It is designed for preparation of Cy5/Cy5.5-labeled compounds and polymers for fluorescence imaging of cellular processes and trafficking. Solid material should be protected from light and stored at -20°C; in solution, store at -80°C for long-term stability.
  • High purity (99.8%) suitable for labeling reactions.
  • Available in small research quantities for synthesis and conjugation workflows.
  • TFA salt form improves solubility in common organic solvents.
  • Compatible with protocols to prepare Cy5/Cy5.5 fluorescent probes for imaging applications.
Catalog No. 50-002-97524 Supplier Medchemexpress LLC Supplier No. HYD1321A10MG
Cy 5 amine TFA is a cyanine fluorescent amine supplied as the trifluoroacetic acid (TFA) salt. It is designed for preparation of Cy5/Cy5.5-labeled compounds and polymers for fluorescence imaging of cellular processes and trafficking. Solid material should be protected from light and stored at -20°C; in solution, store at -80°C for long-term stability.
  • High purity (99.8%) suitable for labeling reactions.
  • Available in small research quantities for synthesis and conjugation workflows.
  • TFA salt form improves solubility in common organic solvents.
  • Compatible with protocols to prepare Cy5/Cy5.5 fluorescent probes for imaging applications.