Learn More
Medchemexpress LLC Cyanine2 | 260430-02-2 | 98.0% | 5 MG
SDP
Catalog No. 501933196
Encompass_Preferred
Cyanines are compounds with two nitrogen atoms linked by an odd number of methene units. The nitrogen atoms are part of heterocyclic units (such as indole, benzoxazol, or benzothiazol). They are characterized by long wavelength, tunable absorption and emission, very high extinction coefficients (up to 300,000 M-1 cm-1), good water solubility, and relatively straightforward synthesis.
  • Long wavelength
  • Tunable absorption and emission
  • Very high extinction coefficients (up to 300,000 M-1 cm-1)
  • Good water solubility
  • Relatively straightforward synthesis
Catalog No. 50-193-3196 Supplier Medchemexpress LLC Supplier No. HYD08275MG
Cyanines are compounds with two nitrogen atoms linked by an odd number of methene units. The nitrogen atoms are part of heterocyclic units (such as indole, benzoxazol, or benzothiazol). They are characterized by long wavelength, tunable absorption and emission, very high extinction coefficients (up to 300,000 M-1 cm-1), good water solubility, and relatively straightforward synthesis.
  • Long wavelength
  • Tunable absorption and emission
  • Very high extinction coefficients (up to 300,000 M-1 cm-1)
  • Good water solubility
  • Relatively straightforward synthesis