Learn More
Apexbio Technology LLC Cy3.5 azide (non-sulfonated), 5mg. Cas: MFCD: NA. used for Click chemistry to label alkyne-modified oligonucleotides
SDP
Supplier:  Apexbio Technology LLC A81125
Encompass_Preferred
Cy3.5 azide is a solid material dye ready for the use in Click Chemistry reaction. Most derivatives of non-sulfonated cyanines have low aqueous solubility except for hydrochlorides of hydrazides and amines Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5009