Learn More
Apexbio Technology LLC Cy3.5 hydrazide (non-sulfonated), 5mg. Cas: N/A MFCD: NA. carbonyl-reactive dye
SDP
Supplier:  Apexbio Technology LLC A82665
Encompass_Preferred
Cy3.5 hydrazide is a near infrared fluorophore that can react with various carbonyl groups in biomolecules which are generated after oxidative stress or deamination of proteins. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5103