Learn More
Apexbio Technology LLC Cy3.5 maleimide (non-sulfonated), 25mg. Cas: N/A MFCD: NA. Labeling of cysteine residues, as well as other thiolated molecules
SDP
Catalog No. 501905096
Encompass_Preferred
Cy3.5 maleimide can be used for selective labeling of free sulfhydryl groups (e.g. cysteine residues in peptides, proteins and oligonucleotides). Before labeling, cystines should be prepared with reductant such as TCEP (tris-carboxyethylphosphine). Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5096 Supplier Apexbio Technology LLC Supplier No. A826425
Cy3.5 maleimide can be used for selective labeling of free sulfhydryl groups (e.g. cysteine residues in peptides, proteins and oligonucleotides). Before labeling, cystines should be prepared with reductant such as TCEP (tris-carboxyethylphosphine). Other sizes are available. Please inqury us for quote.