Learn More
Apexbio Technology LLC Cy3.5 NHS ester (non-sulfonated), 5mg. Cas: MFCD: NA. Labeling of amino-groups in biomolecules.
SDP
Catalog No. 501904990
Encompass_Preferred
Cyanine3.5 NHS ester is a dye for labeling of oligonucleotides, peptides and proteins which contain amino-groups. For fluorescence imaging and other fluorescence-based biochemical analysis, it has been used to label biological molecules too. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4990 Supplier Apexbio Technology LLC Supplier No. A81015
Cyanine3.5 NHS ester is a dye for labeling of oligonucleotides, peptides and proteins which contain amino-groups. For fluorescence imaging and other fluorescence-based biochemical analysis, it has been used to label biological molecules too. Other sizes are available. Please inqury us for quote.