Learn More
Medchemexpress LLC HY-D0822 5mg Medchemexpress, CY3 CAS:146368-13-0 Purity:>98%
SDP
Catalog No. 501933191
Encompass_Preferred
Medchemexpress, HY-D0822 5mg CY3 CAS:146368-13-0 Cy3 (Sulfo-Cyanine3) is an orange-fluorescent label for protein and nucleic acid (λ ex =554, λ em =568). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3191 Supplier Medchemexpress LLC Supplier No. HYD08225MG
Medchemexpress, HY-D0822 5mg CY3 CAS:146368-13-0 Cy3 (Sulfo-Cyanine3) is an orange-fluorescent label for protein and nucleic acid (λ ex =554, λ em =568). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.