Learn More
Apexbio Technology LLC Cy3 alkyne (non-sulfonated), 5mg. Cas: MFCD: NA. used for Click chemistry to label any molecule bearing azide group.
SDP
Supplier:  Apexbio Technology LLC A81305
Encompass_Preferred
Cy3 alkyne is a water-insoluble dye ready for the use in Click Chemistry reaction. With this bright and photostable reagent, molecules bearing azide group can be attached by it. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5041