Learn More
Apexbio Technology LLC Cy3 azide , 25mg. Cas: MFCD: MFCD31580114. used for Click chemistry to label alkyne-modified oligonucleotides
SDP
Catalog No. 501905034
Encompass_Preferred
Sulfo-Cy3 azide is a sulfonated, hydrophilic and highly water soluble dye for Click Chemistry. For labeling of various molecules in aqueous phase for brightness and photostability of fluorophore, sulfo-Cy3 azide is an ideal choice. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5034 Supplier Apexbio Technology LLC Supplier No. A812725
Sulfo-Cy3 azide is a sulfonated, hydrophilic and highly water soluble dye for Click Chemistry. For labeling of various molecules in aqueous phase for brightness and photostability of fluorophore, sulfo-Cy3 azide is an ideal choice. Other sizes are available. Please inqury us for quote.