Learn More
Apexbio Technology LLC Cy3 azide (non-sulfonated), 25mg. Cas: MFCD: NA. used for Click chemistry to label alkyne-modified oligonucleotides
SDP
Supplier:  Apexbio Technology LLC A811125
Encompass_Preferred
Cy3 azide is a non-sulfonated dye ready for the use in Click Chemistry reaction which can be detected by various fluorometers, imagers, and microscopes. The reagent can be detected by naked eye on gels because of inherently high extinction coefficient. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5006