Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cy3-cisplatin | 85.0% | 10 MG
SDP

Catalog No. 5000297534
Encompass_Preferred

Cy3-Cisplatin is a fluorescent conjugate linking a Cyanine 3 dye to cisplatin for imaging, tracking, and drug delivery research. Supplied as an orange to red solid, the compound shows ~85% purity and is unstable in solution, so freshly prepared solutions are recommended. Store at -20°C and protect from light.

  • Fluorescent conjugate for imaging and tracking
  • Suitable for drug delivery and fluorescent labeling
  • Supplied as an orange to red solid
  • Purity approximately 85.0%
  • Store at -20°C and protect from light
  • Unstable in solution; prepare fresh solutions before use

Catalog No. 50-002-97534 Supplier Medchemexpress LLC Supplier No. HYD245210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cy3-Cisplatin is a fluorescent conjugate linking a Cyanine 3 dye to cisplatin for imaging, tracking, and drug delivery research. Supplied as an orange to red solid, the compound shows ~85% purity and is unstable in solution, so freshly prepared solutions are recommended. Store at -20°C and protect from light.

  • Fluorescent conjugate for imaging and tracking
  • Suitable for drug delivery and fluorescent labeling
  • Supplied as an orange to red solid
  • Purity approximately 85.0%
  • Store at -20°C and protect from light
  • Unstable in solution; prepare fresh solutions before use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.