Learn More
Abcam Cy3 Conjugation Kit (Fast) - Lightning-Link, 1MG
Catalog No. 502590108
Encompass_Preferred
Cy3 conjugation / labeling in < 20 mins with 30 secs hands-on time using Cy3 Conjugation Kit (Fast) - Lightning-Link ab188287. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-259-0108 Supplier Abcam Supplier No. AB1882871MG
Cy3 conjugation / labeling in < 20 mins with 30 secs hands-on time using Cy3 Conjugation Kit (Fast) - Lightning-Link ab188287. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement