Learn More
Abcam Cy3 Conjugation Kit (Fast) - Lightning-Link, 100UG
Catalog No. 502590109
Encompass_Preferred
Cy3 conjugation / labeling in < 20 mins with 30 secs hands-on time using Cy3 Conjugation Kit (Fast) - Lightning-Link ab188287. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-259-0109 Supplier Abcam Supplier No. AB188287100UG
Cy3 conjugation / labeling in < 20 mins with 30 secs hands-on time using Cy3 Conjugation Kit (Fast) - Lightning-Link ab188287. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement