Learn More
Abcam Cy3 Conjugation Kit (Fast) - Lightning-Link, 3 x 100UG
Catalog No. 502590107
Encompass_Preferred
Cy3 conjugation / labeling in < 20 mins with 30 secs hands-on time using Cy3 Conjugation Kit (Fast) - Lightning-Link ab188287. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-259-0107 Supplier Abcam Supplier No. AB1882873X100UG
Cy3 conjugation / labeling in < 20 mins with 30 secs hands-on time using Cy3 Conjugation Kit (Fast) - Lightning-Link ab188287. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement