Learn More
Apexbio Technology LLC Cy3 carboxylic acid (non-sulfonated), 5mg
SDP
Supplier:  Apexbio Technology LLC A81325
Encompass_Preferred
Cy3 carboxylic acid is a non-activated fluorescence dye. Most derivatives of non-sulfonated cyanines have low aqueous solubility except for hydrochlorides of hydrazides and amines. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5044