Learn More
Medchemexpress LLC Cy3-dCTP | 2365531-97-9 | 98.70% | 1156.71 | 5 MG
SDP
Catalog No. 5000431241
Encompass_Preferred
Cy3-dCTP is a directly fluorescently labeled deoxyribonucleotide, where Cy3 is a cyanine fluorescent dye. It is used for direct enzymatic labeling of DNA and cDNA, being incorporated into the extending DNA strand during processes such as reverse transcription, PCR, nick translation, or random primer labeling with the aid of DNA polymerases.
  • Directly fluorescently labeled deoxyribonucleotide
  • Cyanine fluorescent dye
  • Used for direct enzymatic labeling of DNA and cDNA
  • Incorporated by DNA polymerases during reverse transcription, PCR, nick translation, or random primer labeling
Catalog No. 50-004-31241 Supplier Medchemexpress LLC Supplier No. HYD17255MG
Cy3-dCTP is a directly fluorescently labeled deoxyribonucleotide, where Cy3 is a cyanine fluorescent dye. It is used for direct enzymatic labeling of DNA and cDNA, being incorporated into the extending DNA strand during processes such as reverse transcription, PCR, nick translation, or random primer labeling with the aid of DNA polymerases.
  • Directly fluorescently labeled deoxyribonucleotide
  • Cyanine fluorescent dye
  • Used for direct enzymatic labeling of DNA and cDNA
  • Incorporated by DNA polymerases during reverse transcription, PCR, nick translation, or random primer labeling