Learn More
Apexbio Technology LLC Cy3-dUTP, 25ul (1mM). Cas: MFCD: NA. Direct enzymatic labeling of DNA/cDNA e.g. by PCR and Nick Translation
SDP
Catalog No. 501905495
Encompass_Preferred
Cy3-dUTP can be used for direct enzymatic labeling of DNA/cDNA by PCR with Taq polymerase, Nick Translation with DNAse I/ DNA Polymerase I, cDNA labeling and 3-end labeling Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5495 Supplier Apexbio Technology LLC Supplier No. B82020.025
Cy3-dUTP can be used for direct enzymatic labeling of DNA/cDNA by PCR with Taq polymerase, Nick Translation with DNAse I/ DNA Polymerase I, cDNA labeling and 3-end labeling Other sizes are available. Please inqury us for quote.