Learn More
Apexbio Technology LLC Cy3 Goat Anti-Mouse IgG (H L) Antibody 1ml
SDP
Catalog No. 503107103
Encompass_Preferred
Anti-mouse secondary antibodies are affinity-purified antibodies specifically recognizing mouse immunoglobulins used for detecting sorting or purifying their respective targets Secondary antibodies offer additional functionalities enabling users to apply various detection systems (such as HRP AP fluorescence) They can also enhance sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody Typically secondary antibodies are generated by immunizing a host animal with pooled immunoglobulins from the target species These antibodies can be further purified and modified (e g immunoaffinity chromatography antibody fragmentation labeling conjugation) to produce highly-specific reagents
Catalog No. 50-310-7103 Supplier Apexbio Technology LLC Supplier No. K12071
Anti-mouse secondary antibodies are affinity-purified antibodies specifically recognizing mouse immunoglobulins used for detecting sorting or purifying their respective targets Secondary antibodies offer additional functionalities enabling users to apply various detection systems (such as HRP AP fluorescence) They can also enhance sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody Typically secondary antibodies are generated by immunizing a host animal with pooled immunoglobulins from the target species These antibodies can be further purified and modified (e g immunoaffinity chromatography antibody fragmentation labeling conjugation) to produce highly-specific reagents