Learn More
Apexbio Technology LLC Cy3 Goat Anti-Human IgG (H+L) Antibody 2X1ml
SDP
Catalog No. 5000097262
Encompass_Preferred
Cy3 Goat Anti-Human IgG HplusL Antibody is a polyclonal antibody targeting human immunoglobulin G (IgG) heavy and light chains It is designed to bind specifically to human IgG primary antibodies through antigen-antibody interactions Cy3 Goat Anti-Human IgG HplusL Antibody exerts its biological activity primarily by enabling indirect detection of target antigens via fluorescent labeling Based on these properties Cy3 Goat Anti-Human IgG HplusL Antibody holds research potential in immunofluorescence microscopy flow cytometry western blot and enzyme-linked immunosorbent assay (ELISA) applications
Catalog No. 50-000-97262 Supplier Apexbio Technology LLC Supplier No. K12082
Cy3 Goat Anti-Human IgG HplusL Antibody is a polyclonal antibody targeting human immunoglobulin G (IgG) heavy and light chains It is designed to bind specifically to human IgG primary antibodies through antigen-antibody interactions Cy3 Goat Anti-Human IgG HplusL Antibody exerts its biological activity primarily by enabling indirect detection of target antigens via fluorescent labeling Based on these properties Cy3 Goat Anti-Human IgG HplusL Antibody holds research potential in immunofluorescence microscopy flow cytometry western blot and enzyme-linked immunosorbent assay (ELISA) applications