Learn More
Apexbio Technology LLC Cy3 hydrazide (non-sulfonated), 5mg. Cas: MFCD: NA. carbonyl-reactive dye
SDP
Supplier:  Apexbio Technology LLC A82655
Encompass_Preferred
Cy3 hydrazide is a carbonyl-reactive dye, an analog of Cy3 hydrazide. Cyanine3 dyes have been used to label biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5100
 Restricted Item