Learn More
Medchemexpress LLC Cy3 methyltetrazine | 2183473-57-4 | C40H46BF4N7O | 1 MG
SDP
Catalog No. 5000373930
Encompass_Preferred
Cy3 methyltetrazine is a click chemistry reagent featuring methyltetrazine building blocks, demonstrating high reactivity towards cyclooctene. It functions as a tetrazine-modified fluorescent probe for analyzing protein phosphorylation in solutions and living cells. This product is intended for research use only.
  • Click chemistry reagent
  • Highly reactive towards cyclooctene
  • Tetrazine-modified fluorescent probe
  • Analyzes protein phosphorylation in solutions and living cells
  • Solid appearance, brown to reddish brown color
  • High purity of 97.36%
Catalog No. 50-003-73930 Supplier Medchemexpress LLC Supplier No. HY1517761MG
Cy3 methyltetrazine is a click chemistry reagent featuring methyltetrazine building blocks, demonstrating high reactivity towards cyclooctene. It functions as a tetrazine-modified fluorescent probe for analyzing protein phosphorylation in solutions and living cells. This product is intended for research use only.
  • Click chemistry reagent
  • Highly reactive towards cyclooctene
  • Tetrazine-modified fluorescent probe
  • Analyzes protein phosphorylation in solutions and living cells
  • Solid appearance, brown to reddish brown color
  • High purity of 97.36%