Learn More
Medchemexpress LLC Cy3 methyltetrazine | 2183473-57-4 | C40H46BF4N7O | 5 MG
SDP
Catalog No. 5000374932
Encompass_Preferred
Cy3 methyltetrazine (TZ-Cy3) is a click chemistry reagent that incorporates methyltetrazine building blocks, making it highly reactive with cyclooctene. It also functions as a tetrazine-modified fluorescent probe, suitable for analyzing protein phosphorylation in solution and within living cells.
  • Click chemistry reagent
  • Reactive with cyclooctene
  • Functions as a tetrazine-modified fluorescent probe
  • Analyzes protein phosphorylation in solution and within living cells
  • For research use only
  • Solid appearance, brown to reddish brown
  • Emission at 570 nm
  • Excitation at 550 nm
Catalog No. 50-003-74932 Supplier Medchemexpress LLC Supplier No. HY1517765MG
Cy3 methyltetrazine (TZ-Cy3) is a click chemistry reagent that incorporates methyltetrazine building blocks, making it highly reactive with cyclooctene. It also functions as a tetrazine-modified fluorescent probe, suitable for analyzing protein phosphorylation in solution and within living cells.
  • Click chemistry reagent
  • Reactive with cyclooctene
  • Functions as a tetrazine-modified fluorescent probe
  • Analyzes protein phosphorylation in solution and within living cells
  • For research use only
  • Solid appearance, brown to reddish brown
  • Emission at 570 nm
  • Excitation at 550 nm